siRNA Details
| Gene Symbol: Aktip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011926.1 | NP_001011926.1 | 3, 4 | 418 | ||
| XM_006255181.2 | XP_006255243.1 | 3, 4 | 430 | |||
| XM_006255182.2 | XP_006255244.1 | 3, 4 | 408 | |||
| XM_006255183.2 | XP_006255245.1 | 2, 3 | 331 | |||
| XM_006255185.2 | XP_006255247.1 | 3, 4 | 364 | |||
| GenBank mRNA | BC089985.1 | - | 3, 4 | 418 | 7318380 | |

Aktip
Genomic Map