siRNA Details
| Gene Symbol: Tox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106171.1 | NP_001099641.1 | 2 | 276 | ||
| XM_006255187.2 | XP_006255249.1 | 3 | 425 | |||
| XM_006255189.2 | XP_006255251.1 | 3 | 422 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tox3| Gene Symbol: Tox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106171.1 | NP_001099641.1 | 2 | 276 | ||
| XM_006255187.2 | XP_006255249.1 | 3 | 425 | |||
| XM_006255189.2 | XP_006255251.1 | 3 | 422 | |||