siRNA Details
| Gene Symbol: Abcc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199377.1 | NP_955409.1 | 25 | 3516 | ||
| XM_006255241.2 | XP_006255303.1 | 25 | 3657 | |||
| XM_006255242.2 | XP_006255304.1 | 24 | 3454 | |||
| XM_008772375.1 | XP_008770597.1 | 25 | 3680 | |||
| XM_008772376.1 | XP_008770598.1 | 25 | 3677 | |||
| XM_008772377.1 | XP_008770599.1 | 24 | 3591 | |||
| XM_008772378.1 | XP_008770600.1 | 25 | 3681 | |||
| GenBank mRNA | AY188179.1 | - | 25 | 3516 | ||
| AY188180.1 | - | 24 | 3279 | |||

Abcc12
Genomic Map