siRNA Details
| Gene Symbol: RGD1308706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134421.1 | NP_001127893.1 | 2 | 208 | ||
| GenBank mRNA | BC167107.1 | - | 2 | 208 | 7110902 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1308706| Gene Symbol: RGD1308706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134421.1 | NP_001127893.1 | 2 | 208 | ||
| GenBank mRNA | BC167107.1 | - | 2 | 208 | 7110902 | |