siRNA Details
| Gene Symbol: Orc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033690.1 | NP_001028862.1 | 3 | 597 | ||
| XM_006255357.1 | XP_006255419.1 | 2 | 187 | |||
| GenBank mRNA | BC101871.1 | - | 3 | 597 | 5621693 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orc6| Gene Symbol: Orc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033690.1 | NP_001028862.1 | 3 | 597 | ||
| XM_006255357.1 | XP_006255419.1 | 2 | 187 | |||
| GenBank mRNA | BC101871.1 | - | 3 | 597 | 5621693 | |