siRNA Details
| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255413.2 | XP_006255475.1 | 14 | 4445 | ||
| XM_008772544.1 | XP_008770766.1 | 14 | 4463 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp827| Gene Symbol: Zfp827 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006255413.2 | XP_006255475.1 | 14 | 4445 | ||
| XM_008772544.1 | XP_008770766.1 | 14 | 4463 | |||