siRNA Details
| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010948.1 | NP_001010948.1 | 8 | 667 | ||
| GenBank mRNA | AY825929.1 | - | 8 | 667 | ||
| FQ211514.1 | - | 858 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc10a7| Gene Symbol: Slc10a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010948.1 | NP_001010948.1 | 8 | 667 | ||
| GenBank mRNA | AY825929.1 | - | 8 | 667 | ||
| FQ211514.1 | - | 858 | ||||