siRNA Details
Gene Symbol: Tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178330.1 | NP_847900.1 | 9 | 1389 | ||
XM_006255422.2 | XP_006255484.1 | 10 | 1101 | |||
XM_008772475.1 | XP_008770697.1 | 10 | 2000 | |||
GenBank mRNA | AY228474.1 | - | 9 | 1389 | ||
BC089112.1 | - | 9 | 1399 | 7316181 |
