siRNA Details
| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191599.1 | NP_001178528.1 | 10 | 2295 | ||
| XM_006255424.2 | XP_006255486.1 | 10 | 2331 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prmt10| Gene Symbol: Prmt10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191599.1 | NP_001178528.1 | 10 | 2295 | ||
| XM_006255424.2 | XP_006255486.1 | 10 | 2331 | |||