siRNA Details
| Gene Symbol: Pgrmc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021766.1 | NP_068534.1 | 1 | 388 | ||
| GenBank mRNA | AF163321.1 | - | 1 | 351 | ||
| AJ005837.1 | - | 1 | 365 | |||
| BC062073.1 | - | 1 | 354 | 5599449 | ||
| U63315.1 | - | 1 | 388 | |||

Pgrmc1
Genomic Map