siRNA Details
| Gene Symbol: Hsf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106177.1 | NP_001099647.1 | 11 | 1169 | ||
| XM_006255453.2 | XP_006255515.1 | 12 | 1827 | |||
| XM_006255454.2 | XP_006255516.1 | 12 | 1737 | |||
| XM_006255455.2 | XP_006255517.1 | 11 | 1718 | |||

Hsf4
Genomic Map