siRNA Details
| Gene Symbol: Tppp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009639.1 | NP_001009639.1 | 3, 4 | 446 | ||
| XM_006255464.2 | XP_006255526.1 | 3, 4 | 476 | |||
| GenBank mRNA | BC087585.1 | - | 3, 4 | 446 | 7317980 | |
| FQ213895.1 | - | 2, 3 | 588 | |||
| FQ214516.1 | - | 3, 4 | 460 | |||
| FQ214740.1 | - | 3, 4 | 450 | |||
| FQ216797.1 | - | 3, 4 | 471 | |||
| FQ217207.1 | - | 3, 4 | 447 | |||

Tppp3
Genomic Map