siRNA Details
| Gene Symbol: Zdhhc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039099.1 | NP_001034188.1 | 5 | 633 | ||
| XM_008772485.1 | XP_008770707.1 | 5 | 2409 | |||
| XM_008772486.1 | XP_008770708.1 | 5 | 2408 | |||
| GenBank mRNA | AY886520.1 | - | 5 | 633 | ||
| BC160816.1 | - | 4 | 552 | |||

Zdhhc1
Genomic Map