siRNA Details
| Gene Symbol: Fam65a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100660.1 | NP_001094130.1 | 13 | 2360 | ||
| XM_006255470.2 | XP_006255532.1 | 13 | 2709 | |||
| XM_006255471.2 | XP_006255533.1 | 13 | 2821 | |||
| XM_006255472.2 | XP_006255534.1 | 13 | 2810 | |||
| XM_006255473.2 | XP_006255535.1 | 13 | 2710 | |||
| XM_006255474.2 | XP_006255536.1 | 13 | 2709 | |||
| XM_006255475.2 | XP_006255537.1 | 13 | 2373 | |||
| GenBank mRNA | AY724537.1 | - | 5 | 1589 | ||
| BC099098.1 | - | 3 | 1327 | |||

Fam65a
Genomic Map