siRNA Details
| Gene Symbol: Enkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106180.1 | NP_001099650.1 | 7 | 1262 | ||
| XM_006255476.2 | XP_006255538.1 | 8 | 1042 | |||
| XM_006255477.2 | XP_006255539.1 | 8 | 1092 | |||
| XM_006255478.2 | XP_006255540.1 | 8 | 1026 | |||

Enkd1
Genomic Map