siRNA Details
| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011928.1 | NP_001011928.1 | 11 | 1382 | ||
| XM_008772487.1 | XP_008770709.1 | 11 | 1679 | |||
| GenBank mRNA | BC088195.1 | - | 11 | 1382 | 7374282 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dpep2| Gene Symbol: Dpep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011928.1 | NP_001011928.1 | 11 | 1382 | ||
| XM_008772487.1 | XP_008770709.1 | 11 | 1679 | |||
| GenBank mRNA | BC088195.1 | - | 11 | 1382 | 7374282 | |