siRNA Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106182.2 | NP_001099652.1 | 2 | 462 | ||
| GenBank mRNA | BC168966.1 | - | 2 | 465 | 7124140 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog8| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106182.2 | NP_001099652.1 | 2 | 462 | ||
| GenBank mRNA | BC168966.1 | - | 2 | 465 | 7124140 | |