siRNA Details
| Gene Symbol: Nob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199086.1 | NP_954517.1 | 7, 8 | 834 | ||
| GenBank mRNA | AY341886.1 | - | 7, 8 | 834 | ||
| BC126076.1 | - | 7, 8 | 842 | 8366716 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nob1| Gene Symbol: Nob1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199086.1 | NP_954517.1 | 7, 8 | 834 | ||
| GenBank mRNA | AY341886.1 | - | 7, 8 | 834 | ||
| BC126076.1 | - | 7, 8 | 842 | 8366716 | ||