siRNA Details
| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013891.1 | NP_001013913.1 | 3 | 452 | ||
| XM_006255579.2 | XP_006255641.1 | 4 | 562 | |||
| GenBank mRNA | BC089962.1 | - | 3 | 452 | 7314335 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Txnl4b| Gene Symbol: Txnl4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013891.1 | NP_001013913.1 | 3 | 452 | ||
| XM_006255579.2 | XP_006255641.1 | 4 | 562 | |||
| GenBank mRNA | BC089962.1 | - | 3 | 452 | 7314335 | |