siRNA Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005381.2 | NP_001005381.2 | 11 | 1277 | ||
| XM_008772490.1 | XP_008770712.1 | 1063 | ||||
| GenBank mRNA | AY303540.1 | - | 1295 | |||
| BC079094.1 | - | 11 | 1248 | 7123989 | ||
| Gene Symbol: Ddx19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005895.2 | NP_001005895.2 | 11 | 1353 | ||
| XM_008772534.1 | XP_008770756.1 | 11 | 1267 | |||
| GenBank mRNA | AY751756.1 | - | 11 | 1295 | ||

Ddx19a
Genomic Map