siRNA Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100517.1 | NP_001093987.1 | 11 | 1493 | ||
| XM_006255580.2 | XP_006255642.1 | 11 | 1731 | |||
| XM_006255581.2 | XP_006255643.1 | 12 | 1588 | |||
| XM_006255582.2 | XP_006255644.1 | 11 | 1727 | |||
| XM_006255583.2 | XP_006255645.1 | 11 | 1483 | |||

Aars
Genomic Map