siRNA Details
| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163273.1 | NP_001156745.1 | 7 | 772 | ||
| XM_006255684.2 | XP_006255746.1 | 5 | 2114 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmip| Gene Symbol: Cmip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163273.1 | NP_001156745.1 | 7 | 772 | ||
| XM_006255684.2 | XP_006255746.1 | 5 | 2114 | |||