siRNA Details
| Gene Symbol: Zc3h18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_201416.1 | NP_958819.1 | 2 | 664 | ||
| XM_006255758.2 | XP_006255820.1 | 3 | 1327 | |||
| XM_006255759.2 | XP_006255821.1 | 3 | 1328 | |||
| XM_006255761.2 | XP_006255823.1 | 3 | 838 | |||
| XM_008772603.1 | XP_008770825.1 | 3 | 746 | |||
| GenBank mRNA | AY389807.1 | - | 2 | 664 | ||

Zc3h18
Genomic Map