siRNA Details
| Gene Symbol: Aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013061.1 | NP_001013079.1 | 1, 2 | 75 | ||
| GenBank mRNA | FQ218973.1 | - | 102 | |||
| FQ228571.1 | - | 1, 2 | 93 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aprt| Gene Symbol: Aprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013061.1 | NP_001013079.1 | 1, 2 | 75 | ||
| GenBank mRNA | FQ218973.1 | - | 102 | |||
| FQ228571.1 | - | 1, 2 | 93 | |||