siRNA Details
| Gene Symbol: Sult5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106194.1 | NP_001099664.1 | 5 | 614 | ||
| NM_001201369.1 | NP_001188298.1 | 6 | 763 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sult5a1| Gene Symbol: Sult5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106194.1 | NP_001099664.1 | 5 | 614 | ||
| NM_001201369.1 | NP_001188298.1 | 6 | 763 | |||