siRNA Details
| Gene Symbol: Urb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135708.1 | NP_001129180.1 | 3 | 2227 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Urb2| Gene Symbol: Urb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135708.1 | NP_001129180.1 | 3 | 2227 | ||