siRNA Details
| Gene Symbol: Ttc13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136162.1 | NP_001129634.1 | 18, 19 | 2128 | ||
| XM_006255802.2 | XP_006255864.1 | 20, 21 | 2296 | |||
| XM_006255803.2 | XP_006255865.1 | 20, 21 | 2293 | |||
| XM_006255805.2 | XP_006255867.1 | 18, 19 | 2134 | |||
| GenBank mRNA | AY724473.1 | - | 8, 9 | 765 | ||
| BC098840.1 | - | 4, 5 | 412 | |||
| BC105871.1 | - | 10, 11 | 1113 | |||

Ttc13
Genomic Map