siRNA Details
| Gene Symbol: Arv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106197.1 | NP_001099667.1 | 4, 5 | 714 | ||
| XM_006255806.2 | XP_006255868.1 | 4, 5 | 672 | |||
| XM_008772606.1 | XP_008770828.1 | 4, 5 | 625 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arv1| Gene Symbol: Arv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106197.1 | NP_001099667.1 | 4, 5 | 714 | ||
| XM_006255806.2 | XP_006255868.1 | 4, 5 | 672 | |||
| XM_008772606.1 | XP_008770828.1 | 4, 5 | 625 | |||