siRNA Details
| Gene Symbol: Pgbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106198.1 | NP_001099668.1 | 3 | 791 | ||
| XM_006255807.2 | XP_006255869.1 | 3 | 878 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgbd5| Gene Symbol: Pgbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106198.1 | NP_001099668.1 | 3 | 791 | ||
| XM_006255807.2 | XP_006255869.1 | 3 | 878 | |||