siRNA Details
Gene Symbol: RGD1562218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034919.1 | NP_001030091.1 | 2, 3 | 421 | ||
GenBank mRNA | BC105747.1 | - | 2, 3 | 421 | 5597965 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034919.1 | NP_001030091.1 | 2, 3 | 421 | ||
GenBank mRNA | BC105747.1 | - | 2, 3 | 421 | 5597965 |