siRNA Details
| Gene Symbol: Sprtn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106199.1 | NP_001099669.1 | 4 | 1158 | ||
| XM_006255808.2 | XP_006255870.1 | 5 | 1550 | |||
| XM_006255809.2 | XP_006255871.1 | 5 | 1307 | |||
| XM_006255810.2 | XP_006255872.1 | 5 | 1333 | |||

Sprtn
Genomic Map