siRNA Details
| Gene Symbol: Habp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001505.1 | NP_001001505.1 | 3 | 151 | ||
| XM_006231631.2 | XP_006231693.1 | 3 | 199 | |||
| GenBank mRNA | AB177406.1 | - | 3 | 151 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Habp2| Gene Symbol: Habp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001505.1 | NP_001001505.1 | 3 | 151 | ||
| XM_006231631.2 | XP_006231693.1 | 3 | 199 | |||
| GenBank mRNA | AB177406.1 | - | 3 | 151 | ||