siRNA Details
| Gene Symbol: Dclre1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106201.1 | NP_001099671.1 | 7 | 2822 | ||
| XM_006231634.1 | XP_006231696.1 | 8 | 3498 | |||
| XM_006231635.2 | XP_006231697.1 | 8 | 3436 | |||
| XM_008760523.1 | XP_008758745.1 | 8 | 3498 | |||

Dclre1a
Genomic Map