siRNA Details
| Gene Symbol: Tdrd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106202.1 | NP_001099672.1 | 6 | 875 | ||
| XM_006231642.2 | XP_006231704.1 | 7 | 1138 | |||
| XM_008760524.1 | XP_008758746.1 | 7 | 945 | |||
| XM_008760525.1 | XP_008758747.1 | 7 | 910 | |||
| XM_008760526.1 | XP_008758748.1 | 6 | 1075 | |||
| XM_008760528.1 | XP_008758750.1 | 6 | 704 | |||
| XM_008760529.1 | XP_008758751.1 | 6 | 982 | |||
| XM_008760530.1 | XP_008758752.1 | 7 | 1138 | |||
| XM_008760531.1 | XP_008758753.1 | 7 | 1138 | |||

Tdrd1
Genomic Map