siRNA Details
| Gene Symbol: Afap1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006231655.2 | XP_006231717.1 | 12 | 1729 | ||
| XM_008760580.1 | XP_008758802.1 | 13 | 1814 | |||
| XM_008760581.1 | XP_008758803.1 | 12 | 1729 | |||
| GenBank mRNA | AB568258.1 | - | 10 | 1198 | ||

Afap1l2
Genomic Map