siRNA Details
| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC085922.1 | - | 2, 3 | 198 | ||
| EF055485.1 | - | 2, 3 | 113 | |||
| EF055488.1 | - | 2, 3 | 113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1311558| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC085922.1 | - | 2, 3 | 198 | ||
| EF055485.1 | - | 2, 3 | 113 | |||
| EF055488.1 | - | 2, 3 | 113 | |||