siRNA Details
| Gene Symbol: RGD1311558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008760532.1 | XP_008758754.1 | 10 | 1092 | ||
| XM_008760533.1 | XP_008758755.1 | 10 | 1109 | |||
| GenBank mRNA | BC085922.1 | - | 12 | 1255 | ||
| CB751427.1 | - | 2 | 197 | |||
| EF055485.1 | - | 13 | 1265 | |||
| EF055488.1 | - | 13 | 1265 | |||

RGD1311558
Genomic Map