siRNA Details
| Gene Symbol: Sh3glb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011929.1 | NP_001011929.1 | 6 | 900 | ||
| XM_006233431.2 | XP_006233493.1 | 8 | 1026 | |||
| XM_006233433.2 | XP_006233495.1 | 8 | 976 | |||
| XM_006233435.2 | XP_006233497.1 | 6 | 912 | |||
| GenBank mRNA | BC079085.1 | - | 6 | 900 | 7123144 | |
| FQ216102.1 | - | 2 | 114 | |||
| FQ234013.1 | - | 6 | 623 | |||

Sh3glb1
Genomic Map