siRNA Details
| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134736.1 | NP_001128208.1 | 7 | 828 | ||
| GenBank mRNA | BC168204.1 | - | 7 | 828 | 7121248 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr63| Gene Symbol: Wdr63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134736.1 | NP_001128208.1 | 7 | 828 | ||
| GenBank mRNA | BC168204.1 | - | 7 | 828 | 7121248 | |