siRNA Details
| Gene Symbol: Ccdc170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758712.1 | XP_008756934.1 | 2369 | |||
| XM_008758717.1 | XP_008756939.1 | 709 | ||||
| XM_008758718.1 | XP_008756940.1 | 709 | ||||
| XM_214760.8 | XP_214760.4 | 2370 | ||||

Ccdc170
Genomic Map