siRNA Details
| Gene Symbol: Ccdc170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758717.1 | XP_008756939.1 | 1417 | |||
| XM_008758719.1 | XP_008756941.1 | 1216 | ||||
| XM_214760.8 | XP_214760.4 | 3078 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc170| Gene Symbol: Ccdc170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008758717.1 | XP_008756939.1 | 1417 | |||
| XM_008758719.1 | XP_008756941.1 | 1216 | ||||
| XM_214760.8 | XP_214760.4 | 3078 | ||||