siRNA Details
| Gene Symbol: Nox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004216.1 | NP_001004216.1 | 4 | 331 | ||
| GenBank mRNA | AY573239.1 | - | 4 | 331 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nox3| Gene Symbol: Nox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004216.1 | NP_001004216.1 | 4 | 331 | ||
| GenBank mRNA | AY573239.1 | - | 4 | 331 | ||