siRNA Details
| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106209.1 | NP_001099679.1 | 6 | 991 | ||
| XM_006227918.2 | XP_006227980.1 | 8 | 1894 | |||
| XM_008758750.1 | XP_008756972.1 | 5 | 1143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

T| Gene Symbol: T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106209.1 | NP_001099679.1 | 6 | 991 | ||
| XM_006227918.2 | XP_006227980.1 | 8 | 1894 | |||
| XM_008758750.1 | XP_008756972.1 | 5 | 1143 | |||