siRNA Details
| Gene Symbol: Olr387 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000109.1 | NP_001000109.1 | 1 | 896 | ||
| Gene Symbol: Olr331 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000760.1 | NP_001000760.1 | 1 | 905 | ||

Olr387
Genomic Map