siRNA Details
| Gene Symbol: B3gnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106211.1 | NP_001099681.1 | 2 | 108 | ||
| XM_006229745.2 | XP_006229807.1 | 2 | 663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B3gnt6| Gene Symbol: B3gnt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106211.1 | NP_001099681.1 | 2 | 108 | ||
| XM_006229745.2 | XP_006229807.1 | 2 | 663 | |||