siRNA Details
Gene Symbol: RGD1562747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106212.1 | NP_001099682.1 | 3 | 692 | ||
GenBank mRNA | FQ213024.1 | - | 3 | 667 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD1562747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106212.1 | NP_001099682.1 | 3 | 692 | ||
GenBank mRNA | FQ213024.1 | - | 3 | 667 |