siRNA Details
| Gene Symbol: Fpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106216.1 | NP_001099686.1 | 1 | 658 | ||
| XM_006227988.2 | XP_006228050.1 | 2 | 845 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fpr1| Gene Symbol: Fpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106216.1 | NP_001099686.1 | 1 | 658 | ||
| XM_006227988.2 | XP_006228050.1 | 2 | 845 | |||
Set Membership: |
|