siRNA Details
| Gene Symbol: Stx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025638.2 | NP_001020809.2 | 2 | 1526 | ||
| XM_008758601.1 | XP_008756823.1 | 2 | 1574 | |||
| GenBank mRNA | BC099159.1 | - | 1 | 1193 | 7381603 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx11| Gene Symbol: Stx11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025638.2 | NP_001020809.2 | 2 | 1526 | ||
| XM_008758601.1 | XP_008756823.1 | 2 | 1574 | |||
| GenBank mRNA | BC099159.1 | - | 1 | 1193 | 7381603 | |