siRNA Details
| Gene Symbol: Fuca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004218.1 | NP_001004218.1 | 4 | 887 | ||
| GenBank mRNA | BC078933.1 | - | 4 | 887 | 7107565 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fuca2| Gene Symbol: Fuca2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004218.1 | NP_001004218.1 | 4 | 887 | ||
| GenBank mRNA | BC078933.1 | - | 4 | 887 | 7107565 | |