siRNA Details
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134425.1 | NP_001127897.1 | 4 | 442 | ||
| XM_006227642.2 | XP_006227704.1 | 4 | 641 | |||
| XM_008758602.1 | XP_008756824.1 | 4 | 638 | |||
| GenBank mRNA | BC167098.1 | - | 4 | 442 | 7319120 | |
| FQ228880.1 | - | 4 | 514 | |||
| FQ229550.1 | - | 4 | 459 | |||
| FQ229983.1 | - | 4 | 458 | |||

Aig1
Genomic Map